Assay Details
Target Gene Details
Entrez Gene ID: | 56208 |
Gene Name: | beclin 1, autophagy related |
Gene Aliases: |
4921513J16Rik, 5430417M23Rik, Atg6 |
Location: |
Chr.11:101288262-101302288 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Becn1 | NM_019584.3 | 9 | 1003 | NP_062530.2 |
| AF077302.2 | 8 | 865 | AAC68654.2 | |
| AK028851.1 | 7 | 2445 | ||
| AK031501.1 | 8 | 1630 | ||
| AK050329.1 | 9 | 1004 | BAC34192.1 | |
| AK077392.1 | 9 | 1090 | ||
| AK083800.1 | 9 | 993 | BAC39021.1 | |
| AK151531.1 | 9 | 970 | ||
| AK215649.1 | ||||
| BC005770.1 | 9 | 984 | AAH05770.1 |
More Information
Set Membership: |
|

Genomic Map