Assay Details
Target Gene Details
Entrez Gene ID: | 72947 |
Gene Name: | 5-phosphohydroxy-L-lysine phospholyase |
Gene Aliases: |
2900006B13Rik, Agxt2l2 |
Location: |
Chr.11:51583983-51603269 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phykpl | NM_028398.2 | NP_082674.1 | ||
| AK013489.1 | ||||
| AK085984.1 | BAC39584.1 | |||
| AK156866.1 | ||||
| AK160557.1 | BAE35872.1 | |||
| AK217003.1 | ||||
| BC024461.1 | AAH24461.1 |
More Information
Set Membership: |
|

Genomic Map