Assay Details
Target Gene Details
Entrez Gene ID: | 11932 |
Gene Name: | ATPase, Na+/K+ transporting, beta 2 polypeptide |
Gene Aliases: |
Amog, Atpb-2 |
Location: |
Chr.11:69599750-69605960 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp1b2 | NM_013415.5 | NP_038201.1 | ||
| AK141786.1 | BAE24833.1 | |||
| AK158321.1 | ||||
| BC042467.1 | AAH42467.1 | |||
| BC058763.1 | AAH58763.1 | |||
| X16645.1 | CAA34638.1 |
More Information
Set Membership: |
|

Genomic Map