Assay Details
Target Gene Details
Entrez Gene ID: | 210004 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1 |
Gene Aliases: |
6030413G23Rik |
Location: |
Chr.11:121616197-121673160 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3gntl1 | NM_178664.5 | 12 | 1165 | NP_848779.2 |
| AK031361.1 | 12 | 1154 | BAC27366.1 | |
| AK156318.1 | 12 | 1143 | BAE33671.1 | |
| BC022739.1 | 6 | 477 | AAH22739.1 | |
| BC088993.1 | 12 | 1030 | AAH88993.1 | |
| BC125626.1 | 12 | 1138 | ||
| BC132193.1 | 12 | 1138 | ||
| BC145166.1 | 12 | 1138 |
More Information
Set Membership: |
|

Genomic Map