Assay Details
Target Gene Details
Entrez Gene ID: | 73324 |
Gene Name: | clathrin heavy chain linker domain containing 1 |
Gene Aliases: |
1700034F02Rik |
Location: |
Chr.11:29547950-29578360 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clhc1 | NM_001163521.1 | 1 | 44 | NP_001156993.1 |
| NM_028504.1 | 1 | 44 | NP_082780.1 | |
| AK006591.1 | 1 | 46 | BAB24663.1 | |
| BC137683.1 | ||||
| BC144828.1 |
Target Gene Details
Entrez Gene ID: | 78294 |
Gene Name: | ribosomal protein S27A |
Gene Aliases: |
0610006J14Rik, Uba52, Ubb, Ubc |
Location: |
Chr.11:29545842-29548040 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps27a | NM_001033865.1 | 1 | 48 | NP_001029037.1 |
| NM_024277.2 | 1 | 48 | NP_077239.1 |
More Information
Set Membership: |
|

Genomic Map