Assay Details
Target Gene Details
Entrez Gene ID: | 11298 |
Gene Name: | arylalkylamine N-acetyltransferase |
Gene Aliases: |
AA-NAT, Nat-2, Nat4, Snat |
Location: |
Chr.11:116591721-116597680 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aanat | NM_009591.3 | NP_033721.1 | ||
| NR_033223.1 | ||||
| AB013358.1 | BAA31526.1 | |||
| AF004108.1 | AAD09408.1 | |||
| AF004109.1 | AAD09409.1 | |||
| AF004110.1 | AAD09410.1 | |||
| AF004111.1 | AAD09411.1 | |||
| AY131262.1 | ||||
| BC116967.1 | ||||
| BC119139.1 |
More Information
Set Membership: |
|

Genomic Map