Assay Details
Target Gene Details
Entrez Gene ID: | 69684 |
Gene Name: | alanyl-tRNA synthetase domain containing 1 |
Gene Aliases: |
1110069E20Rik, 2310044P18Rik, AA589600, AlaX, AlaXp-II |
Location: |
Chr.11:101406665-101417433 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aarsd1 | NM_144829.1 | NP_659078.1 | ||
| AK078016.1 | BAC37098.1 | |||
| AK136332.1 | BAE22937.1 | |||
| AK168281.1 | BAE40227.1 | |||
| BC005711.1 | AAH05711.1 |
More Information
Set Membership: |
|

Genomic Map