Assay Details
Target Gene Details
Entrez Gene ID: | 67131 |
Gene Name: | acyl-Coenzyme A binding domain containing 4 |
Gene Aliases: |
2010009P05Rik, 2010015A21Rik, AI849317 |
Location: |
Chr.11:103101688-103112199 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acbd4 | NM_025988.2 | NP_080264.1 | ||
| AK008170.1 | ||||
| AK008207.1 | BAB25532.1 | |||
| AK008243.1 | BAC25208.1 | |||
| AK079604.1 | BAC37699.1 | |||
| AK165381.1 | 1 | 98 | ||
| BC048371.1 | AAH48371.1 | |||
| BI734653.1 |
More Information
Set Membership: |
|

Genomic Map