Assay Details
Target Gene Details
Entrez Gene ID: | 380732 |
Gene Name: | mast cell immunoglobulin like receptor 1 |
Gene Aliases: |
Allergin-1, Gm885, Mca32 |
Location: |
Chr.11:106750652-106768794 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Milr1 | NM_001033435.3 | NP_001028607.1 | ||
| NM_001271372.1 | NP_001258301.1 | |||
| NM_001271373.1 | NP_001258302.1 | |||
| NM_001271374.1 | NP_001258303.1 | |||
| NM_001271375.1 | NP_001258304.1 | |||
| NR_073165.1 | ||||
| AB542953.1 | ||||
| AK136488.1 | BAE23003.1 | |||
| AK137500.1 | BAE23382.1 | |||
| AK169925.1 | BAE41463.1 | |||
| AK170947.1 | BAE42134.1 | |||
| AK171381.1 | BAE42422.1 | |||
| BC139224.1 | ||||
| BC139225.1 | ||||
| BC145580.1 | ||||
| BC145582.1 |
Target Gene Details
Entrez Gene ID: | 50776 |
Gene Name: | polymerase (DNA directed), gamma 2, accessory subunit |
Gene Aliases: |
- |
Location: |
Chr.11:106754821-106780010 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polg2 |
More Information
Set Membership: |
|

Genomic Map