Assay Details
Target Gene Details
Entrez Gene ID: | 14694 |
Gene Name: | guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 |
Gene Aliases: |
AL033335, GB-like, Gnb2-rs1, Rack1, p205 |
Location: |
Chr.11:48800360-48806241 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnb2l1 | NM_008143.3 | NP_032169.1 | ||
AK002493.1 | BAB22141.1 | |||
AK012242.1 | BAB28114.1 | |||
AK017772.1 | BAB30920.1 | |||
AK051226.1 | BAC34564.1 | |||
AK159797.1 | BAE35378.1 | |||
AK160528.1 | BAE35846.1 | |||
AK160739.1 | BAE35980.1 | |||
AK161234.1 | BAE36257.1 | |||
AK166417.1 | BAE38762.1 | |||
AK166786.1 | BAE39017.1 | |||
AK166800.1 | BAE39028.1 | |||
AK166868.1 | 2 | 3751 | ||
AK166889.1 | BAE39095.1 | |||
AK166945.1 | BAE39133.1 | |||
AK168086.1 | BAE40059.1 | |||
AK168096.1 | BAE40068.1 | |||
AK168196.1 | BAE40156.1 | |||
AK168349.1 | BAE40286.1 | |||
AK169612.1 | ||||
AK186349.1 | ||||
BC046760.1 | AAH46760.1 | |||
D29802.1 | BAA06185.1 | |||
X75313.1 | CAA53062.1 |
