Assay Details
Target Gene Details
Entrez Gene ID: | 14710 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2 |
Gene Aliases: |
AV096488 |
Location: |
Chr.11:95842219-95845734 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gngt2 | NM_001038664.2 | NP_001033753.2 | ||
| NM_001284393.1 | NP_001271322.1 | |||
| NM_023121.2 | NP_075610.1 | |||
| AA028742.1 | ||||
| AA423571.1 | ||||
| AK010554.1 | 1 | 107 | BAB27023.1 | |
| BC061005.1 | AAH61005.1 | |||
| BE847751.1 | ||||
| BY117112.1 | ||||
| W35863.1 |
More Information
Set Membership: |
|

Genomic Map