Assay Details
Target Gene Details
Entrez Gene ID: | 66610 |
Gene Name: | ABI gene family, member 3 |
Gene Aliases: |
2210414K06Rik, AI987680, NESH |
Location: |
Chr.11:95830072-95842476 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abi3 | NM_001163464.1 | NP_001156936.1 | ||
| NM_025659.4 | 1 | 165 | NP_079935.1 | |
| AK037144.1 | BAC29720.1 |
Target Gene Details
Entrez Gene ID: | 14710 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2 |
Gene Aliases: |
AV096488 |
Location: |
Chr.11:95842219-95845734 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gngt2 | NM_001284393.1 | NP_001271322.1 | ||
| NM_023121.2 | NP_075610.1 | |||
| AA423571.1 | ||||
| BE847751.1 | ||||
| BY117112.1 |
More Information
Set Membership: |
|

Genomic Map