Assay Details
Target Gene Details
Entrez Gene ID: | 216859 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 |
Gene Aliases: |
Centb1 |
Location: |
Chr.11:69881567-69895539 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acap1 | NM_153788.3 | 14 | 1403 | NP_722483.2 |
| AK029819.1 | 4 | 1556 | ||
| AK133473.1 | 10 | 2074 | ||
| AK138736.1 | 14 | 1396 | ||
| AK154449.1 | 14 | 1406 | BAE32594.1 | |
| AK155001.1 | 14 | 1424 | ||
| BC031462.1 | 14 | 1403 | AAH31462.1 | |
| BC037481.1 | 9 | 1030 | AAH37481.1 |

Genomic Map