Assay Details
Target Gene Details
Entrez Gene ID: | 216859 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 |
Gene Aliases: |
Centb1 |
Location: |
Chr.11:69881567-69895539 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acap1 | NM_153788.3 | 17 | 1795 | NP_722483.2 |
| AK029819.1 | 5 | 2569 | ||
| AK133473.1 | 12 | 2993 | ||
| AK138736.1 | 17 | 1788 | ||
| AK154449.1 | 17 | 1798 | BAE32594.1 | |
| AK155001.1 | 17 | 1816 | ||
| BC031462.1 | 17 | 1795 | AAH31462.1 | |
| BC037481.1 | 12 | 1422 | AAH37481.1 |

Genomic Map