Assay Details
Target Gene Details
Entrez Gene ID: | 276852 |
Gene Name: | DNA segment, Chr 11, Wayne State University 47, expressed |
Gene Aliases: |
- |
Location: |
Chr.11:113684400-113727444 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D11Wsu47e | NM_177777.5 | NP_808445.2 | ||
| AK017508.1 | BAB30780.1 | |||
| AK139389.1 | BAE23989.1 | |||
| AK171307.1 | BAE42383.1 | |||
| BC026908.1 | AAH26908.2 | |||
| BC048942.1 | AAH48942.1 |

Genomic Map