Assay Details
Target Gene Details
Entrez Gene ID: | 28081 |
Gene Name: | family with sequence similarity 104, member A |
Gene Aliases: |
D11Wsu99e |
Location: |
Chr.11:113661319-113684186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam104a | NM_138598.5 | NP_613064.2 | ||
| AK009763.2 | BAB26487.1 | |||
| AK017659.1 | BAB30858.1 | |||
| AK028386.1 | BAC25923.1 | |||
| AK087540.1 | BAC39919.1 | |||
| AK145119.1 | ||||
| AK158680.1 | BAE34607.1 | |||
| AK158872.1 | BAE34705.1 | |||
| AK159228.1 | BAE34914.1 | |||
| AK159659.1 | BAE35268.1 | |||
| AV572367.1 | ||||
| BC060965.1 | AAH60965.1 | |||
| BC089526.1 | AAH89526.1 |

Genomic Map