Assay Details
Target Gene Details
Entrez Gene ID: | 217207 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 8 |
Gene Aliases: |
Ddx8, mDEAH6, mKIAA4096 |
Location: |
Chr.11:101732956-101767358 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx8 | NM_144831.2 | 13 | 1983 | NP_659080.2 |
| AK078552.1 | 13 | 1922 | ||
| AK142226.1 | 13 | 1917 | BAE24985.1 | |
| AK170616.1 | 13 | 1912 | ||
| AK172600.1 | 13 | 1929 | ||
| AK220485.1 | 13 | 1960 | BAD90286.1 | |
| BC132445.1 | 13 | 1967 |
More Information
Set Membership: |
|

Genomic Map