Assay Details
Target Gene Details
Entrez Gene ID: | 69131 |
Gene Name: | cyclin-dependent kinase 12 |
Gene Aliases: |
1810022J16Rik, AI646528, Crk7, Crkrs, D11Ertd752e, Pksc |
Location: |
Chr.11:98202326-98281101 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk12 | NM_001109626.1 | 12 | 3209 | NP_001103096.1 |
| NM_001109628.1 | 12 | 3209 | NP_001103098.1 | |
| NM_026952.2 | 12 | 3209 | NP_081228.2 | |
| AK049376.1 | 12 | 2855 | ||
| AK129237.1 | 12 | 2348 | BAC98047.1 | |
| AK137734.1 | 10 | 1079 | ||
| AY072295.1 | 12 | 3200 | AAL69526.1 | |
| BC116645.1 | 12 | 3192 |
More Information
Set Membership: |
|

Genomic Map