Assay Details
Target Gene Details
Entrez Gene ID: | 69131 |
Gene Name: | cyclin-dependent kinase 12 |
Gene Aliases: |
1810022J16Rik, AI646528, Crk7, Crkrs, D11Ertd752e, Pksc |
Location: |
Chr.11:98202326-98281101 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk12 | NM_001109626.1 | 6 | 2558 | NP_001103096.1 |
| NM_001109628.1 | 6 | 2558 | NP_001103098.1 | |
| NM_026952.2 | 6 | 2558 | NP_081228.2 | |
| AK047916.1 | 2 | 2656 | ||
| AK049376.1 | 6 | 2204 | ||
| AK129237.1 | 6 | 1697 | BAC98047.1 | |
| AK137734.1 | 4 | 428 | ||
| AK164881.1 | 6 | 2419 | BAE37951.1 | |
| AY072295.1 | 6 | 2549 | AAL69526.1 | |
| BC116645.1 | 6 | 2541 |
More Information
Set Membership: |
|

Genomic Map