Assay Details
Target Gene Details
Entrez Gene ID: | 276770 |
Gene Name: | eukaryotic translation initiation factor 5A |
Gene Aliases: |
AA410058, D19Wsu54e, Eif4d, Eif5a1, eIF-4D, eIF-5A, eIF-5A-1, eIF-5A1 |
Location: |
Chr.11:69916712-69921958 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
More Information
Set Membership: |
|

Genomic Map