Assay Details
Target Gene Details
Entrez Gene ID: | 12957 |
Gene Name: | crystallin, beta A1 |
Gene Aliases: |
BA3/A1, Cryb |
Location: |
Chr.11:77718732-77725781 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cryba1 | NM_009965.2 | NP_034095.1 | ||
| AJ239052.1 | CAB52418.1 | |||
| BC107342.2 | AAI07343.1 | |||
| M21473.1 | AAA51628.1 |

Genomic Map