Assay Details
Target Gene Details
Entrez Gene ID: | 69306 |
Gene Name: | EF-hand calcium binding domain 9 |
Gene Aliases: |
1700007I06Rik, 4930403C08Rik |
Location: |
Chr.11:32522733-32533271 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab9 | NM_027031.3 | 1 | 134 | NP_081307.2 |
| AK005716.1 | 1 | 141 | BAB24202.1 | |
| AK015057.1 | 1 | 125 | BAB29694.1 | |
| AK161300.1 | 1 | 125 | BAE36307.1 | |
| BC087911.1 | 1 | 134 | AAH87911.1 |
More Information
Set Membership: |
|

Genomic Map