Assay Details
Target Gene Details
Entrez Gene ID: | 13169 |
Gene Name: | drebrin-like |
Gene Aliases: |
Abp1, SH3P7, mAbp1 |
Location: |
Chr.11:5788483-5800980 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbnl | NM_001146308.1 | NP_001139780.1 | ||
| NM_001146309.1 | NP_001139781.1 | |||
| NM_013810.3 | NP_038838.1 | |||
| AK046073.1 | BAC32592.1 | |||
| AK053796.1 | BAC35528.1 | |||
| AK078082.1 | BAC37118.1 | |||
| AK134136.1 | BAE22028.1 | |||
| AK134487.1 | BAE22160.1 | |||
| AK140752.1 | BAE24466.1 | |||
| AK142818.1 | BAE25201.1 | |||
| AK146920.1 | BAE27532.1 | |||
| AK151096.1 | BAE30108.1 | |||
| AK152110.1 | ||||
| AK152204.1 | BAE31033.1 | |||
| AK153072.1 | ||||
| AK153389.1 | BAE31953.1 | |||
| AK159984.1 | BAE35535.1 | |||
| AK161973.1 | ||||
| AK167956.1 | ||||
| AK168898.1 | BAE40714.1 | |||
| AK172471.1 | BAE43024.1 | |||
| AK199448.1 | ||||
| AK203166.1 | ||||
| AY098595.1 | AAM28340.1 | |||
| BC046430.1 | AAH46430.1 | |||
| BY293844.1 | ||||
| U58884.1 | AAC52640.1 |
More Information
Set Membership: |
|

Genomic Map