Assay Details
Target Gene Details
Entrez Gene ID: | 13169 |
Gene Name: | drebrin-like |
Gene Aliases: |
Abp1, SH3P7, mAbp1 |
Location: |
Chr.11:5788483-5800980 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbnl | NM_001146308.1 | 6 | 592 | NP_001139780.1 |
| NM_001146309.1 | 6 | 592 | NP_001139781.1 | |
| NM_013810.3 | 6 | 592 | NP_038838.1 | |
| AK046073.1 | 6 | 572 | BAC32592.1 | |
| AK053796.1 | 6 | 588 | BAC35528.1 | |
| AK078082.1 | 6 | 559 | BAC37118.1 | |
| AK134136.1 | 6 | 559 | BAE22028.1 | |
| AK134487.1 | 6 | 559 | BAE22160.1 | |
| AK140752.1 | 6 | 559 | BAE24466.1 | |
| AK142818.1 | 6 | 542 | BAE25201.1 | |
| AK146920.1 | 6 | 559 | BAE27532.1 | |
| AK151096.1 | 6 | 572 | BAE30108.1 | |
| AK152110.1 | 6 | 557 | ||
| AK152204.1 | 6 | 572 | BAE31033.1 | |
| AK153072.1 | 6 | 571 | ||
| AK153389.1 | 6 | 546 | BAE31953.1 | |
| AK159984.1 | 6 | 569 | BAE35535.1 | |
| AK161973.1 | 6 | 572 | ||
| AK167956.1 | 6 | 531 | ||
| AK168898.1 | 6 | 569 | BAE40714.1 | |
| AK172471.1 | 6 | 549 | BAE43024.1 | |
| AY098595.1 | 6 | 515 | AAM28340.1 | |
| BC046430.1 | 6 | 557 | AAH46430.1 | |
| U58884.1 | 6 | 545 | AAC52640.1 |
More Information
Set Membership: |
|

Genomic Map