Assay Details
Target Gene Details
Entrez Gene ID: | 216616 |
Gene Name: | epidermal growth factor-containing fibulin-like extracellular matrix protein 1 |
Gene Aliases: |
- |
Location: |
Chr.11:28853205-28926743 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efemp1 | NM_146015.2 | NP_666127.2 | ||
| AK077302.1 | BAC36738.1 | |||
| AY251056.1 | AAP79577.1 |
More Information
Set Membership: |
|

Genomic Map