Assay Details
Target Gene Details
Entrez Gene ID: | 216616 |
Gene Name: | epidermal growth factor-containing fibulin-like extracellular matrix protein 1 |
Gene Aliases: |
- |
Location: |
Chr.11:28853205-28926743 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efemp1 | NM_146015.2 | NP_666127.2 | ||
| AK077302.1 | BAC36738.1 | |||
| AK162532.1 | BAE36959.1 | |||
| AY185605.1 | AAO37642.1 | |||
| AY251056.1 | AAP79577.1 | |||
| BC023060.1 | AAH23060.1 | |||
| BC031184.1 | AAH31184.1 |
More Information
Set Membership: |
|

Genomic Map