Assay Details
Target Gene Details
Entrez Gene ID: | 53413 |
Gene Name: | exocyst complex component 7 |
Gene Aliases: |
Exo70, sec70 |
Location: |
Chr.11:116287998-116306749 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exoc7 | NM_001162872.1 | NP_001156344.1 | ||
| NM_016857.2 | NP_058553.2 | |||
| AF014461.1 | AAB69345.1 | |||
| AK087601.1 | BAC39941.1 | |||
| AK140450.1 | BAE24393.1 | |||
| AK162868.1 | ||||
| AK167769.1 | BAE39802.1 | |||
| AK169928.1 | BAE41466.1 | |||
| AK173094.1 | BAD32372.1 | |||
| BC028927.1 | AAH28927.1 | |||
| BC053710.1 | AAH53710.1 |
More Information
Set Membership: |
|

Genomic Map