Assay Details
Target Gene Details
Entrez Gene ID: | 83429 |
Gene Name: | cystinosis, nephropathic |
Gene Aliases: |
AI195360, AW049661 |
Location: |
Chr.11:73183133-73199029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctns | NM_031251.4 | NP_112541.1 | ||
| AJ250670.1 | CAC19455.1 | |||
| AK054467.1 | BAC35789.1 | |||
| AK078511.1 | BAC37316.1 | |||
| AK079295.1 | BAC37598.1 | |||
| AK085517.1 | ||||
| AK137143.1 | BAE23249.1 | |||
| AK170790.1 | BAE42030.1 | |||
| AK170828.1 | BAE42058.1 | |||
| BC005479.1 | AAH05479.1 |
More Information
Set Membership: |
|

Genomic Map