Assay Details
Target Gene Details
Entrez Gene ID: | 171212 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10 |
Gene Aliases: |
AU018154, C330012K04Rik, GalNAc-T10, GalNAc-T9, Galnt9, ppGaNTase |
Location: |
Chr.11:57622407-57787501 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt10 | NM_134189.2 | NP_598950.2 | ||
| AK033515.1 | BAC28334.1 | |||
| AK131155.1 | BAD21405.1 | |||
| AK143714.1 | ||||
| AK159088.1 | BAE34806.1 | |||
| AK159362.1 | BAE35020.1 | |||
| AK162424.1 | ||||
| BC016585.1 | AAH16585.1 | |||
| BC060617.1 | AAH60617.1 |
More Information
Set Membership: |
|

Genomic Map