Assay Details
Target Gene Details
Entrez Gene ID: | 216963 |
Gene Name: | G protein-coupled receptor kinase-interactor 1 |
Gene Aliases: |
Cat-1, p95Cat |
Location: |
Chr.11:77493412-77507775 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Git1 | NM_001004144.1 | NP_001004144.1 | ||
| AK050701.1 | BAE20687.1 | |||
| BC079870.1 | AAH79870.1 |
More Information
Set Membership: |
|

Genomic Map