Assay Details
Target Gene Details
Entrez Gene ID: | 15218 |
Gene Name: | forkhead box N1 |
Gene Aliases: |
D11Bhm185e, Fkh19, HFH-11, Hfh11, Whn, nu, nude |
Location: |
Chr.11:78357577-78386922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxn1 | NM_001277290.1 | NP_001264219.1 | ||
| NM_008238.2 | NP_032264.1 | |||
| BC108980.1 | AAI08981.1 | |||
| BC108981.1 | AAI08982.1 | |||
| CJ056441.1 | ||||
| X81593.1 | CAA57279.1 |

Genomic Map