Assay Details
Target Gene Details
Entrez Gene ID: | 93896 |
Gene Name: | glucagon-like peptide 2 receptor |
Gene Aliases: |
9530092J08Rik, GLP-2 |
Location: |
Chr.11:67663299-67771153 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glp2r | NM_175681.3 | 6 | 822 | NP_783612.2 |
| AF166265.1 | 2 | 62 | AAF89584.1 | |
| AY255567.1 | AAO85079.1 | |||
| AY605231.1 | 6 | 541 | AAT46060.1 | |
| BC044746.2 | 6 | 822 | AAH44746.1 |
More Information
Set Membership: |
|

Genomic Map