Assay Details
Target Gene Details
Entrez Gene ID: | 13207 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 5 |
Gene Aliases: |
2600009A06Rik, G17P1, HUMP68, Hlr1, p68 |
Location: |
Chr.11:106780355-106789268 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx5 | NM_007840.3 | NP_031866.2 | ||
| AK044792.1 | ||||
| AK076018.1 | ||||
| AK088887.1 | BAC40633.1 | |||
| AK129022.1 | ||||
| AK132779.1 | BAE21355.1 | |||
| AK135173.1 | BAE22450.1 | |||
| AK166045.1 | BAE38540.1 | |||
| AK166698.1 | BAE38954.1 | |||
| BC062916.1 | ||||
| BC086320.1 | AAH86320.1 | |||
| BC108369.1 | AAI08370.1 | |||
| BC129873.1 | ||||
| BC129874.1 | ||||
| X65627.1 | CAA46581.1 |

Genomic Map