Assay Details
Target Gene Details
Entrez Gene ID: | 215512 |
Gene Name: | family with sequence similarity 117, member A |
Gene Aliases: |
5730593F17Rik, AW548096 |
Location: |
Chr.11:95337018-95381872 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam117a | NM_172543.4 | 5 | 726 | NP_766131.2 |
| AK077747.1 | 5 | 727 | BAC36990.1 | |
| AK143632.1 | 5 | 727 | ||
| AK165378.1 | 5 | 727 | BAE38154.1 | |
| AK167417.1 | 5 | 727 | BAE39505.1 | |
| BC055690.1 | 5 | 698 | AAH55690.1 |

Genomic Map