Assay Details
Target Gene Details
Entrez Gene ID: | 14457 |
Gene Name: | growth arrest specific 7 |
Gene Aliases: |
AW124766, B230343A10Rik, Gas7-cb |
Location: |
Chr.11:67455024-67688992 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas7 | NM_001109657.3 | NP_001103127.1 | ||
| NM_001277079.1 | NP_001264008.1 | |||
| NM_001277080.1 | NP_001264009.1 | |||
| NM_008088.5 | NP_032114.2 | |||
| AF133184.1 | AAF15397.1 | |||
| AK017394.1 | ||||
| AK076926.1 | ||||
| AK152149.1 | BAE30985.1 | |||
| AK154463.1 | BAE32603.1 | |||
| AK172943.1 | BAD32221.1 | |||
| AK200726.1 | ||||
| BC120528.1 | ||||
| BC120530.1 | ||||
| BU053145.1 | ||||
| U19860.1 | AAA85259.1 |
Target Gene Details
Entrez Gene ID: | 93896 |
Gene Name: | glucagon-like peptide 2 receptor |
Gene Aliases: |
9530092J08Rik, GLP-2 |
Location: |
Chr.11:67663299-67771153 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glp2r |
More Information
Set Membership: |
|

Genomic Map