Assay Details
Target Gene Details
Entrez Gene ID: | 13649 |
Gene Name: | epidermal growth factor receptor |
Gene Aliases: |
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2 |
Location: |
Chr.11:16752203-16913907 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egfr | NM_007912.4 | 3 | 571 | NP_031938.1 |
| NM_207655.2 | 3 | 571 | NP_997538.1 | |
| AF124513.1 | 3 | 505 | AAD44149.1 | |
| AF275367.1 | 3 | 514 | AAG24386.1 | |
| AK004883.1 | 3 | 572 | BAB23641.1 | |
| AK004911.1 | 3 | 546 | BAB23662.1 | |
| AK004944.1 | 3 | 565 | BAB23688.1 | |
| AK134262.1 | 3 | 545 | ||
| BC023729.1 | 3 | 542 | AAH23729.1 | |
| L06864.1 | 3 | 257 | AAA53029.1 | |
| U03425.1 | 3 | 341 | AAA17899.1 | |
| X59698.1 | 3 | 450 | CAA42219.1 | |
| X78987.1 | 3 | 350 | CAA55587.1 |
More Information
Set Membership: |
|

Genomic Map