Assay Details
Target Gene Details
Entrez Gene ID: | 70383 |
Gene Name: | cytochrome c oxidase assembly protein 10 |
Gene Aliases: |
2410004F01Rik, AU042636 |
Location: |
Chr.11:63962627-64079472 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox10 | NM_178379.3 | NP_848466.1 | ||
| AK010385.1 | ||||
| AK149999.1 | BAE29227.1 | |||
| AK154668.1 | BAE32754.1 | |||
| AK156865.1 | BAE33878.1 | |||
| AK171661.1 | BAE42594.1 | |||
| AK172718.1 | BAE43143.1 | |||
| AK178239.1 | ||||
| AK179799.1 | ||||
| AK189355.1 | ||||
| AK196412.1 | ||||
| AK199666.1 | ||||
| AK206048.1 | ||||
| AK206430.1 | ||||
| AK207639.1 | ||||
| AK210222.1 | ||||
| AK212782.1 | ||||
| AK212812.1 | ||||
| AK213695.1 | ||||
| AK218286.1 | ||||
| BC038046.1 | AAH38046.1 | |||
| BY078593.1 |
More Information
Set Membership: |
|

Genomic Map