Assay Details
Target Gene Details
Entrez Gene ID: | 70383 |
Gene Name: | cytochrome c oxidase assembly protein 10 |
Gene Aliases: |
2410004F01Rik, AU042636 |
Location: |
Chr.11:63962627-64079472 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox10 | NM_178379.3 | NP_848466.1 | ||
| AK010385.1 | ||||
| AK149999.1 | BAE29227.1 | |||
| AK154668.1 | BAE32754.1 | |||
| AK171661.1 | BAE42594.1 | |||
| AK178210.1 | ||||
| AK184676.1 | ||||
| AK186700.1 | ||||
| BC038046.1 | AAH38046.1 |
More Information
Set Membership: |
|

Genomic Map