Assay Details
Target Gene Details
Entrez Gene ID: | 216820 |
Gene Name: | dehydrogenase/reductase (SDR family) member 7B |
Gene Aliases: |
BC003479, C79874, C80074, PexRAP |
Location: |
Chr.11:60830631-60858423 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhrs7b | NM_001172112.1 | NP_001165583.1 | ||
| NM_145428.2 | NP_663403.1 | |||
| AK052209.1 | BAC34887.1 | |||
| AK139084.1 | BAE23884.1 | |||
| AK144057.1 | BAE25674.1 | |||
| AK205316.1 | ||||
| BC003479.1 | AAH03479.1 | |||
| JX196872.1 |
More Information
Set Membership: |
|

Genomic Map