Assay Details
Target Gene Details
Entrez Gene ID: | 13866 |
Gene Name: | v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) |
Gene Aliases: |
Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023 |
Location: |
Chr.11:98412484-98437716 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb2 | NM_001003817.1 | NP_001003817.1 | ||
| AK031099.1 | ||||
| AK083669.1 | ||||
| AK129487.1 | BAC98297.1 | |||
| AK142296.1 | ||||
| AK215635.1 | ||||
| BC046811.1 | AAH46811.1 | |||
| BC053078.1 | AAH53078.1 | |||
| CF728071.1 |
More Information
Set Membership: |
|

Genomic Map