Assay Details
Target Gene Details
Entrez Gene ID: | 52670 |
Gene Name: | cleavage and polyadenylation specific factor 4-like |
Gene Aliases: |
0610010C04Rik, 1500000C01Rik, D11Ertd636e |
Location: |
Chr.11:113696078-113711425 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf4l | NM_001164532.1 | NP_001158004.1 | ||
| NM_026682.2 | NP_080958.2 | |||
| NM_029794.2 | NP_084070.1 | |||
| AK002449.1 | BAE43147.1 | |||
| AK005072.1 | BAB23796.1 | |||
| AK161074.1 | 4 | 432 | BAE36185.1 | |
| BC004603.1 | 1 | 96 | AAH04603.1 | |
| BC016190.1 | AAH16190.1 | |||
| DV648079.1 | ||||
| DV659633.1 |
Target Gene Details
Entrez Gene ID: | 276852 |
Gene Name: | DNA segment, Chr 11, Wayne State University 47, expressed |
Gene Aliases: |
- |
Location: |
Chr.11:113684400-113727444 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D11Wsu47e | AK139389.1 | BAE23989.1 |
More Information
Set Membership: |
|

Genomic Map