Assay Details
Target Gene Details
Entrez Gene ID: | 20918 |
Gene Name: | eukaryotic translation initiation factor 1 |
Gene Aliases: |
Sui1-rs1 |
Location: |
Chr.11:100319994-100322096 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1 | NM_011508.1 | NP_035638.1 | ||
| AF129888.1 | AAD31266.1 | |||
| AK148032.1 | BAE28300.1 | |||
| AK150813.1 | BAE29876.1 | |||
| AK151070.1 | BAE30086.1 | |||
| AK151575.1 | BAE30515.1 | |||
| AK152194.1 | BAE31024.1 | |||
| AK169763.1 | BAE41352.1 | |||
| BC003463.1 | AAH03463.1 | |||
| BC010791.1 | AAH10791.1 | |||
| BC081429.1 | AAH81429.1 | |||
| BC096656.1 | AAH96656.1 | |||
| BC096691.1 | AAH96691.1 | |||
| Z50159.1 | CAA90519.1 |
More Information
Set Membership: |
|

Genomic Map