Assay Details
Target Gene Details
Entrez Gene ID: | 78943 |
Gene Name: | endoplasmic reticulum (ER) to nucleus signalling 1 |
Gene Aliases: |
9030414B18Rik, AI225830, C85377, Ire1a, Ire1alpha, Ire1p |
Location: |
Chr.11:106397620-106487796 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ern1 | NM_023913.2 | NP_076402.1 | ||
| AB031332.1 | BAB20901.1 | |||
| AK018505.1 | BAB31243.1 | |||
| AK080709.1 | ||||
| AK170988.1 | BAE42162.1 |
More Information
Set Membership: |
|

Genomic Map