Assay Details
Target Gene Details
Entrez Gene ID: | 24083 |
Gene Name: | N-acetyltransferase domain containing 1 |
Gene Aliases: |
AI256713, F3-2, Gm16515, Gtlf3b |
Location: |
Chr.11:60902246-60913792 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Natd1 | NM_025294.5 | 2 | 221 | NP_079570.1 |
| AK004720.2 | 2 | 220 | BAB23503.1 | |
| AK080453.1 | 2 | 221 | BAC37922.1 | |
| AK089179.1 | 2 | 220 | BAC40780.1 | |
| AK201883.1 | ||||
| BC034332.1 | 2 | 177 | AAH34332.1 | |
| BC038816.1 | 2 | 219 | AAH38816.1 | |
| BY196792.1 | 2 | 221 |

Genomic Map