Assay Details
Target Gene Details
Entrez Gene ID: | 14919 |
Gene Name: | guanylate cyclase 2e |
Gene Aliases: |
GC-E, GC1, ROS-GC1 |
Location: |
Chr.11:69218117-69237358 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy2e | NM_008192.3 | 10 | 2176 | NP_032218.2 |
| AK044282.1 | 10 | 2177 | BAC31853.1 | |
| BI738500.1 | 1 | 127 |
More Information
Set Membership: |
|

Genomic Map