Assay Details
Target Gene Details
Entrez Gene ID: | 14919 |
Gene Name: | guanylate cyclase 2e |
Gene Aliases: |
GC-E, GC1, ROS-GC1 |
Location: |
Chr.11:69218117-69237358 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy2e | NM_008192.3 | 12 | 2477 | NP_032218.2 |
| BI738500.1 | 3 | 429 |
More Information
Set Membership: |
|

Genomic Map