Assay Details
Target Gene Details
Entrez Gene ID: | 216850 |
Gene Name: | KDM1 lysine (K)-specific demethylase 6B |
Gene Aliases: |
1700064E03Rik, BC038313, Jmjd3, mKIAA0346 |
Location: |
Chr.11:69398508-69418148 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm6b | NM_001017426.1 | 9 | 1271 | NP_001017426.1 |
| BC075632.1 | 10 | 1447 | AAH75632.1 | |
| CA945559.1 | 3 | 357 |
More Information
Set Membership: |
|

Genomic Map