Assay Details
Target Gene Details
Entrez Gene ID: | 360013 |
Gene Name: | myosin XVIIIA |
Gene Aliases: |
MAJN, MyoPDZ, MysPDZ |
Location: |
Chr.11:77763251-77865988 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo18a | NM_001291212.1 | 12 | 2243 | NP_001278141.1 |
| NM_001291213.1 | 12 | 2351 | NP_001278142.1 | |
| NM_001291214.1 | 10 | 1198 | NP_001278143.1 | |
| NM_001291215.1 | 13 | 1264 | NP_001278144.1 | |
| NM_011586.3 | 11 | 2207 | NP_035716.1 | |
| AB026497.1 | 11 | 2177 | BAA93660.1 | |
| AK137574.1 | 11 | 1198 | BAE23413.1 | |
| AK147584.1 | 13 | 1213 | BAE28009.1 | |
| AK171342.1 | 10 | 1195 | BAE42402.1 | |
| BC138366.1 | 12 | 2185 |

Genomic Map