Assay Details
Target Gene Details
Entrez Gene ID: | 103844 |
Gene Name: | inhibitor of CDK, cyclin A1 interacting protein 1 |
Gene Aliases: |
AI842396 |
Location: |
Chr.11:70688361-70700206 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inca1 | NM_001252482.1 | 3 | 345 | NP_001239411.1 |
| NM_001252483.1 | 3 | 306 | NP_001239412.1 | |
| NM_001252484.1 | 3 | 236 | NP_001239413.1 | |
| NM_001252485.1 | 4 | 465 | NP_001239414.1 | |
| NM_213729.1 | 3 | 374 | NP_998894.1 | |
| AK139002.1 | 3 | 234 | BAE23857.1 | |
| AY601909.1 | 3 | 374 | AAT09154.1 | |
| AY601910.1 | 3 | 345 | AAT09155.1 | |
| AY601911.1 | 3 | 306 | AAT09156.1 | |
| AY601912.1 | 3 | 236 | AAT09157.1 | |
| BC096427.1 | 4 | 419 | AAH96427.1 | |
| BC115572.1 | 3 | 205 | ||
| BC115573.1 | 3 | 205 |

Genomic Map