Assay Details
Target Gene Details
Entrez Gene ID: | 107732 |
Gene Name: | mitochondrial ribosomal protein L10 |
Gene Aliases: |
0610040E02Rik, C78440, MRP-L8, Rpml8 |
Location: |
Chr.11:97041567-97049213 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl10 | NM_026154.1 | NP_080430.1 | ||
| AK002865.1 | BAB22415.1 | |||
| AK007643.1 | BAB25158.1 | |||
| AK078463.1 | ||||
| AK078499.1 | 1 | 3116 | ||
| AK150774.1 | BAE29840.1 | |||
| AK171028.1 | BAE42195.1 | |||
| BC016219.1 | AAH16219.1 |
More Information
Set Membership: |
|

Genomic Map